2-Formylimidazol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1H-Imidazole-2-carbaldehyde, Imidazole-2-carboxaldehyde
M-Clarity™ Qualitätsstufe = MQ100
272000-1G
272000-5G
SIAL272000-1GEA
42.4
EUR
SIAL272000-1G
SIAL272000-5G
2-Formylimidazol, Sigma-Aldrich®
2-Formylimidazol
Formel:
C₄H₄N₂O Molekulargewicht: 96.09 g/mol |
MDL Number:
MFCD00003544 CAS-Nummer: 10111-08-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...