Storage drums, Rotilabo®
Lieferant: Roth Carl
A859.1 A860.1 0489.1 0490.1 0491.1
ROTHA859.1EA 341 EUR
ROTHA859.1 ROTHA860.1 ROTH0489.1 ROTH0490.1 ROTH0491.1
Storage drums, Rotilabo®
Fässer, Fässer, Trommeln
Stackable drum, relatively light and impervious to fluids.
  • Ground and brushed inside and out
  • Drum made of 1,4541 stainless steel
  • Lid with silicone seal and 1,4301 stainless steel tension ring
  • Surfaces in contact with product Ra ≤0,6 µm