Stoppers, rubber, with hole
Lieferant: Roth Carl
Y846.1 Y847.1 Y848.1 Y849.1 Y850.1 Y851.1 Y852.1 Y853.1 Y854.1 Y855.1
ROTHY846.1EA 6.6 EUR
ROTHY846.1 ROTHY847.1 ROTHY848.1 ROTHY849.1 ROTHY850.1 ROTHY851.1 ROTHY852.1 ROTHY853.1 ROTHY854.1 ROTHY855.1
Stoppers, rubber, with hole
Stopfen Gummistopfen
Made of elastic natural rubber. With hole. Hardness 40 Shore A.
  • Silicone-free
  • Temperature stable from −35 to +70 °C
  • Autoclavable