2-Aminoimidazolhemisulfat 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Aminoimidazole sulphate, 1H-imidazol-2-amine sulphate(2:1)
H25999.03
H25999.09
H25999.03EA
41.6
EUR
H25999.03
H25999.09
2-Aminoimidazolhemisulfat 98%
2-Aminoimidazolhemisulfat
Purity: 98%
CAS No.: 1450-93-7
Molecular Formula: C6H10N6.0.5H2SO4
Formula Weight: 215.22
Odour: Odourless
Melting Point: ca 270° dec.
Beilstein Reference: 4726750
MDL No.: MFCD00013162
Formel:
C₆H₁₂N₆O₄S Schmelzpunkt: ∼ 270 °C (dec.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00013162 CAS-Nummer: 1450-93-7 EINECS: 215-918-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...