1,4-Diethinylbenzol 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
p-Diethynylbenzene, 1,4-Diethylnylbenzene
391550050
391550010
ACRO391550050EA
169
EUR
ACRO391550050
ACRO391550010
1,4-Diethinylbenzol 95%
1,4-Diethynylbenzene
Formel:
C₁₀H₆ Schmelzpunkt: 94…98 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00078375 CAS-Nummer: 935-14-8 |
Spezifikation Testergebnisse
Appearance | White to orange to brown Crystalline powder, crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 90 to 96 °C |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...