Methyl-4-phenylbenzoat ≥98%
Lieferant: Thermo Fisher Scientific
A13430.06
A13430.18
A13430.06EA
38.2
EUR
A13430.06
A13430.18
Methyl-4-phenylbenzoat ≥98%
Methyl-4-phenylbenzoat
Purity: 98+%
CAS No.: 720-75-2
Molecular Formula: C14H12O2
Formula Weight: 212.25
Odour: Odourless
Melting Point: 116-119°
Beilstein Reference: 1954046
MDL No.: MFCD00017200
Formel:
C₁₄H₁₂O₂ Schmelzpunkt: 116…119 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00017200 CAS-Nummer: 720-75-2 EINECS: 211-954-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...