Lanthantriacetat Sequihydrat ≥99,9%
Lieferant: Thermo Fisher Scientific
Synonyme:
Lanthanum(III) acetate sesquihydrate
15299.22
15299.36
AB203924.0500
15299.22EA
67.8
EUR
15299.22
15299.36
ABCRAB203924.0500
Lanthantriacetat Sequihydrat ≥99,9%
Lanthantriacetat Sequihydrat
Purity: 99.9%
CAS No.: 25721-92-0
Molecular Formula: La(OOCCH3)3.1.5H2O
Formula Weight: 343.05 (316.03anhy)
Physical Form: Lump/Powder
Sensitivity: Hygroscopic
MDL No.: MFCD00013037
Formel:
C₆H₉LaO₆·1.5H₂O Molekulargewicht: 343.05 g/mol (316.03 anhy) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00013037 CAS-Nummer: 25721-92-0 EINECS: 213-034-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...