(Benzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (BOP) 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate, ((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)tris(dimethylamino)phosphonium hexafluorophosphate(V), BOP reagent, Benzotriazol-1-yloxytris(dimethylamino)-phosphonium hexafluorophosphate
Purity: 98%
CAS No.: 56602-33-6
Molecular Formula: C12H22F6N6OP2
Formula Weight: 442.29
Storage Temperature: 2°C to 8°C
Melting Point: ca 138° dec.
Beilstein Reference: 4287025
UN No.: 1325
MDL No.: MFCD00011948
Formel:
C₁₂H₂₂F₆N₆OP₂ Molekulargewicht: 442.28 g/mol Schmelzpunkt: ∼ 138 °C (Zers.) |
MDL Number:
MFCD00011948 CAS-Nummer: 56602-33-6 EINECS: 260-279-1 ADR: 4.1 II |