3,5-Dichlor-2-hydroxybenzolsulfonsäure Natriumsalz ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: Sodium 3,5-Dichloro-2-hydroxybenzenesulphonate, DHBS

A10732.09 A10732.18 A10732.30
A10732.09EA 57.2 EUR
A10732.09 A10732.18 A10732.30
3,5-Dichlor-2-hydroxybenzolsulfonsäure Natriumsalz ≥99%
3,5-Dichlor-2-hydroxybenzolsulfonsäure Natriumsalz

Purity: 99% CAS No.: 54970-72-8 Molecular Formula: C6H3Cl2NaO4S Formula Weight: 265.05 Melting Point: 300° Beilstein Reference: 3640643

Formel: C₆H₃Cl₂NaO₄S
Molekulargewicht: 265.05 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00009798
CAS-Nummer: 54970-72-8
EINECS: 259-416-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR