D(+)-Glucose-[1-D] (98% D), Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
310816-1G
310816-250MG
SIAL310816-1GEA
314
EUR
SIAL310816-1G
SIAL310816-250MG
D(+)-Glucose-[1-D] (98% D), Sigma-Aldrich®
D(+)-Glucose-[1-D]
Formel:
C₆H₁₁DO₆ |
MDL Number:
MFCD00074868 CAS-Nummer: 106032-61-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...