(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphoniumhexafluorophosphat (PyBOP) ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
PyBOP, ((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)tri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V), 1H-Benzotriazol-1-yloxytri(1-pyrrolidinyl)phosphonium hexafluorophosphate
Purity: 98%
CAS No.: 128625-52-5
Molecular Formula: C18H28F6N6OP2
Formula Weight: 520.40
Storage Temperature: 2°C to 8°C
Melting Point: ca 155° dec.
Beilstein Reference: 3584612
MDL No.: MFCD00077411
Formel:
C₁₈H₂₈F₆N₆OP₂ Molekulargewicht: 520.4 g/mol Schmelzpunkt: ∼ 155 °C (Zers.) |
MDL Number:
MFCD00077411 CAS-Nummer: 128625-52-5 |