Cer(III)perchlorat Hexahydrat, Reagenzqualität
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
11654.18
11654.30
11654.18EA
67.2
EUR
11654.18
11654.30
Cer(III)perchlorat Hexahydrat, Reagenzqualität
Cer(III)perchlorat Hexahydrat
CAS No.: 14017-47-1
Molecular Formula: Ce(ClO4)3.6H2O
Formula Weight: 546.56 (438.47anhy)
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Odour: Odourless
UN No.: 1481
MDL No.: MFCD00150205
Formel:
CeCl₃O₁₂·6H₂O Molekulargewicht: 546.56 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00150205 CAS-Nummer: 14017-47-1 EINECS: 237-836-2 UN-Nummer: 1481 ADR: 5.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...