4,4-Dimethylcyclohexanol
Lieferant: Thermo Scientific
BTBG00026.250
BTBG00026DA
BTBG00026.0010
CHMPBTBG00026.250EA
0
EUR
CHMPBTBG00026.250
MAYBBTBG00026DA
MAYBBTBG00026.0010
4,4-Dimethylcyclohexanol
4,4-Dimethylcyclohexanol
Formel:
C₈H₁₆O |
MDL Number:
MFCD00101954 CAS-Nummer: 932-01-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...