2-Isopropyl-N,N,6-trimethylanilin

Lieferant: Thermo Scientific

SEW05770.1 SEW05770.10 SEW05770.50
MAYBSEW05770.1EA 0 EUR
MAYBSEW05770.1 MAYBSEW05770.10 MAYBSEW05770.50
2-Isopropyl-N,N,6-trimethylanilin
2-Isopropyl-N,N,6-trimethylanilin
Formel: C₁₂H₁₉N
CAS-Nummer: 227199-09-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR