Kaliumbis(trimethylsilyl)amid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
324671-25G
324671-100G
324671-5G
SIAL324671-25GEA
129
EUR
SIAL324671-25G
SIAL324671-100G
SIAL324671-5G
Kaliumbis(trimethylsilyl)amid, Sigma-Aldrich®
Formel:
C₆H₁₈KNSi₂ |
MDL Number:
MFCD00010330 CAS-Nummer: 40949-94-8 EINECS: 424-100-2 UN-Nummer: 3263 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...