Trimethylamin-N-oxid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Trimethylamine N-oxide, Trimethylamine oxide, TMANO

M-Clarity™ quality level = MQ100

317594-1G 317594-5G
SIAL317594-1GEA 88.2 EUR
SIAL317594-1G SIAL317594-5G
Trimethylamin-N-oxid, Sigma-Aldrich®
Trimethylamin-N-oxid
Formel: C₃H₉NO
MDL Number: MFCD00002048
CAS-Nummer: 1184-78-7
EINECS: 214-675-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR