D(+)-Cellobiose, Sigma-Aldrich®

Lieferant: Merck

Synonyme: β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose

M-Clarity™ quality level = MQ200

C7252-100G C7252-1KG C7252-25G C7252-500G
SIALC7252-100GEA 472 EUR
SIALC7252-100G SIALC7252-1KG SIALC7252-25G SIALC7252-500G
D(+)-Cellobiose, Sigma-Aldrich®
D(+)-Cellobiose
Formel: C₁₂H₂₂O₁₁
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00136034
CAS-Nummer: 528-50-7
EINECS: 208-436-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR