Di(2-pyridyl)keton 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Di(pyridin-2-yl)methanone
H55322.03
H55322.06
H55322.14
H55322.03EA
48.1
EUR
H55322.03
H55322.06
H55322.14
Di(2-pyridyl)keton 98%
Di(2-pyridyl)keton
Purity: 99%
CAS No.: 19437-26-4
Molecular Formula: C11H8N2O
Formula Weight: 184.20
Flash Point: 110°(230°F)
Melting Point: 52-55°
Beilstein Reference: 126598
MDL No.: MFCD00006288
Formel:
C₁₁H₈N₂O Molekulargewicht: 184.20 g/mol Schmelzpunkt: 52…55 °C Flammpunkt: 110 °C (230 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00006288 CAS-Nummer: 19437-26-4 EINECS: 243-067-3 |
Spezifikation Testergebnisse
Appearance (Color) | White to pale brown |
Form | Solid |
Assay (unspecified) |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...