4'-(Methylsulfonyl)acetophenon

Lieferant: Thermo Scientific

Synonyme: 4'-(Methylsulphonyl)acetophenone

JFD01520.1 JFD01520EA
MAYBJFD01520.1EA 0 EUR
MAYBJFD01520.1 MAYBJFD01520EA
4'-(Methylsulfonyl)acetophenon
4'-(Methylsulfonyl)acetophenon
Formel: C₉H₁₀O₃S
MDL Number: MFCD00025072
CAS-Nummer: 10297-73-1
EINECS: 233-672-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR