2-Brom-4'-(diethylamino)acetophenon 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-(N,N-Diethylamino)phenacyl bromide
L13337.03
L13337.06
L13337.03EA
81.7
EUR
L13337.03
L13337.06
2-Brom-4'-(diethylamino)acetophenon 98%
2-Brom-4'-(diethylamino)acetophenon
Purity: 98%
CAS No.: 207986-25-2
Molecular Formula: C12H16BrNO
Formula Weight: 270.18
Melting Point: 50-52°
Beilstein Reference: 7921481
Formel:
C₁₂H₁₆BrNO Schmelzpunkt: 50…52 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00151798 CAS-Nummer: 207986-25-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...