Piceatannol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

P0453-25MG P0453-5MG
SIALP0453-25MGEA 850 EUR
SIALP0453-25MG SIALP0453-5MG
Piceatannol, Sigma-Aldrich®
Piceatannol
Formel: C₁₄H₁₂O₄
MDL Number: MFCD00221715
CAS-Nummer: 10083-24-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR