N-Methyl-4-boronobenzolsulfonamid ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Methylsulphamoylbenzeneboronic acid, (4-(N-Methylsulphamoyl)phenyl)boronic acid
H52493.MD
H52493.03
H52493.MDEA
136
EUR
H52493.MD
H52493.03
N-Methyl-4-boronobenzolsulfonamid ≥97%
N-Methyl-4-boronobenzolsulfonamid
Purity: 97%
CAS No.: 226396-31-2
Molecular Formula: C7H10BNO4S
Formula Weight: 215.03
Melting Point: 136-140°
MDL No.: MFCD06659864
Formel:
C₇H₁₀BNO₄S Schmelzpunkt: 136…140 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06659864 CAS-Nummer: 226396-31-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...