trans,trans-Dibenzylidenaceton ≥98%
Lieferant: Thermo Fisher Scientific
A12095.06
A12095.14
A12095.22
A12095.06EA
43.6
EUR
A12095.06
A12095.14
A12095.22
trans,trans-Dibenzylidenaceton ≥98%
trans,trans-Dibenzylidenaceton
Purity: 98+%
CAS No.: 35225-79-7
Molecular Formula: C17H14O
Formula Weight: 234.30
Melting Point: 112-114°
Beilstein Reference: 608434
Merck Reference: 14,3006
MDL No.: MFCD00004790
Formel:
C₁₇H₁₄O Schmelzpunkt: 112…114 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00004790 CAS-Nummer: 35225-79-7 Merck Index: 14,3006 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...