Tris[N,N-bis(trimethylsilyl)amid]gadolinium(III) ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

44359.03 44359.06
44359.03EA 128 EUR
44359.03 44359.06
Tris[N,N-bis(trimethylsilyl)amid]gadolinium(III) ≥98%
Tris[N,N-bis(trimethylsilyl)amid]gadolinium(III)

Purity: 98% CAS No.: 35789-03-8 Molecular Formula: C18H54GdN3Si6 Formula Weight: 638.41 Physical Form: Powder Sensitivity: Moisture Sensitive UN No.: 3396 MDL No.: MFCD03427091

Formel: C₁₈H₅₄GdN₃Si₆
Schmelzpunkt: 161…167 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03427091
CAS-Nummer: 35789-03-8
UN-Nummer: 3396
ADR: 4.3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR