4'-(Phenylethinyl)acetophenon

Lieferant: Thermo Scientific

KM08946.1 KM08946.10 KM08946.50
MAYBKM08946.1EA 0 EUR
MAYBKM08946.1 MAYBKM08946.10 MAYBKM08946.50
4'-(Phenylethinyl)acetophenon
4'-(Phenylethinyl)acetophenon
Formel: C₁₆H₁₂O
MDL Number: MFCD00219774
CAS-Nummer: 1942-31-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR