5-Amino-4-brom-1-methyl-1H-pyrazol
Lieferant: Thermo Scientific
Synonyme:
4-Bromo-1-methyl-1H-pyrazol-5-amine
GK03207.1
GK03207.10
MAYBGK03207.1EA
0
EUR
MAYBGK03207.1
MAYBGK03207.10
5-Amino-4-brom-1-methyl-1H-pyrazol
5-Amino-4-bromo-1-methyl-1H-pyrazole
Formel:
C₄H₆BrN₃ |
MDL Number:
MFCD00068157 CAS-Nummer: 105675-85-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...