2-(Trimethylsilyl)ethoxymethylchlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

238902-100G 238902-1G 238902-25G 238902-5G
SIAL238902-100GEA 1430 EUR
SIAL238902-100G SIAL238902-1G SIAL238902-25G SIAL238902-5G
2-(Trimethylsilyl)ethoxymethylchlorid, Sigma-Aldrich®
2-(Trimethylsilyl)ethoxymethylchlorid
Formel: C₆H₁₅ClOSi
MDL Number: MFCD00009919
CAS-Nummer: 76513-69-4
EINECS: 278-483-4
UN-Nummer: 2920
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR