Ionomycin

Lieferant: Cayman Chemical

10004974-1 10004974-5 10004974-10 10004974-25
CAYM10004974-1EA 59 EUR
CAYM10004974-1 CAYM10004974-5 CAYM10004974-10 CAYM10004974-25
Ionomycin
Ionomycin
Formel: C₄₁H₇₂O₉
Molekulargewicht: 709.02 g/mol
MDL Number: MFCD06798385
CAS-Nummer: 56092-81-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR