N(α)-Boc-D-histidin
Lieferant: Novabiochem (Merck Biosciences)
Synonyme:
Boc-D-His-OH
8.53090.0025
8.53090.0005
04-13-0014-25EA
941
EUR
04-13-0014-25
04-13-0014-5
N(α)-Boc-D-histidin
N(α)-Boc-D-histidin
Formel:
C₁₁H₁₇N₃O₄ Molekulargewicht: 255.3 g/mol |
MDL Number:
MFCD00037851 CAS-Nummer: 50654-94-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...