Hesperidin 97% includes analogeous compounds
Lieferant: Thermo Fisher Scientific
Synonyme:
(S)-5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)chroman-4-one
123460050
123462500
ACRO123460050EA
18.8
EUR
ACRO123460050
ACRO123462500
Hesperidin 97% includes analogeous compounds
Hesperidin
Formel:
C₂₈H₃₄O₁₅ Schmelzpunkt: 258…262 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00075663 CAS-Nummer: 520-26-3 EINECS: 208-288-1 |
Spezifikation Testergebnisse
Appearance | White to light brown Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤4 % |
Heavy metals | ≤20 ppm |
Additional info |
residue by calcination: |
UV spectrophotometry | ≥96.0 % (on dry substance) |
Volatile matter | ≤5.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...