2-(2-Pyrrolidinyl)pyridin 96%
Lieferant: Thermo Fisher Scientific
H50176.03
H50176.MD
H50176.03EA
295
EUR
H50176.03
H50176.MD
2-(2-Pyrrolidinyl)pyridin 96%
2-(2-Pyrrolidinyl)pyridin
Purity: 96%
CAS No.: 77790-61-5
Molecular Formula: C9H12N2
Formula Weight: 148.21
UN No.: 2810
MDL No.: MFCD01691538
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...