Glycin Hydrochlorid (H-Gly-OH.HCl) ≥98%
Lieferant: Thermo Fisher Scientific
Achtung
|
Synonyme:
H-Gly-OH·HCl, Glycine hydrochloride
A12162.22
A12162.36
A12162.22EA
60.1
EUR
A12162.22
A12162.36
Glycin Hydrochlorid (H-Gly-OH.HCl) ≥98%
Glycin Hydrochlorid (H-Gly-OH.HCl)
Purity: 98%
CAS No.: 6000-43-7
Molecular Formula: H2NCH2CO2H.HCl
Formula Weight: 111.53
Odour: Odourless
Melting Point: 175-178°
Beilstein Reference: 3909617
Merck Reference: 14,4491
MDL No.: MFCD00012872
Formel:
C₂H₆ClNO₂ Schmelzpunkt: 175…178 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00012872 CAS-Nummer: 6000-43-7 EINECS: 227-841-8 Merck Index: 14,4491 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...