N-Boc-glycin (Boc-Gly-OH) ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Boc-Gly-OH
A11579.06
A11579.14
A11579.22
A11579.06EA
29.4
EUR
A11579.06
A11579.14
A11579.22
N-Boc-glycin (Boc-Gly-OH) ≥98%
N-Boc-glycin (Boc-Gly-OH)
Purity: 98+%
CAS No.: 4530-20-5
Molecular Formula: (CH3)3COCONHCH2CO2H
Formula Weight: 175.18
Melting Point: 87-90°
Beilstein Reference: 1101514
Formel:
C₇H₁₃NO₄ Molekulargewicht: 175.2 g/mol Schmelzpunkt: 87…90 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00002690 CAS-Nummer: 4530-20-5 EINECS: 224-864-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...