3,3'-Diaminobenzidin Tetrahydrochlorid Hydrat 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
DAB HCl hydrate, 3,3',4,4'-Biphenyltetramine tetrahydrochloride hydrate, DAB tetrahydrochloride hydrate
CAS No.: 868272-85-9
Molecular Formula: C12H14N4.4HCl.xH2O
Formula Weight: 360.11(anhy)
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Grey
Solubility: Soluble at 5%(w/v) in water
MDL No.: MFCD08273058
Widely used peroxidase substrate for immunoblotting and immunohistochemical staining techniques
Formel:
C₁₂H₁₄N₄·4HCl·H₂O Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD08273058 CAS-Nummer: 868272-85-9 EINECS: 231-018-9 |