2'-Desoxycytidin
Lieferant: Thermo Fisher Scientific
Synonyme:
4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, deoxyC
J63062.03
J63062.06
J63062.14
J63062.03EA
36.4
EUR
J63062.03
J63062.06
J63062.14
2'-Desoxycytidin
2'-Desoxycytidin
CAS No.: 951-77-9
Molecular Formula: C9H13N3O4
Formula Weight: 227.22
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline powder
Appearance: White
Melting Point: 209-211°
Beilstein Reference: 87567
MDL No.: MFCD00006547
Formel:
C₉H₁₃N₃O₄ Schmelzpunkt: 209…211 °C |
MDL Number:
MFCD00006547 CAS-Nummer: 951-77-9 EINECS: 213-454-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...