1-Naphthylphosphat Mononatriumsalz Monohydrat ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-Naphthylphosphoric Acid Monosodium Salt Monohydrate, Monosodium 1-Naphthyl Phosphate Monohydrate, Sodium 1-Naphthyl Hydrogen Phosphate Monohydrate, Phosphoric Acid 1-Naphthyl Ester Monosodium Salt Monohydrate
Purity: 97+%
CAS No.: 81012-89-7
Molecular Formula: C10H8NaO4P.H2O
Formula Weight: 264.15 (246.13anhy)
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Solubility: Soluble in water
Beilstein Reference: 1877764
MDL No.: MFCD00150615
Substrate for determination of phosphatases
Formel:
C₁₀H₈NaO₄P·1H₂O Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00150615 CAS-Nummer: 81012-89-7 |