3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Hydrat 99+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine dihydrochloride xhydrate, TMB-HCl hydrate, Dihydrochloride TMB hydrate
Purity: 99+%
CAS No.: 207738-08-7
Molecular Formula: C16H20N2.2HCl.xH2O
Formula Weight: 313.27(anhy)
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Sensitivity: Moisture and Light Sensitive
Appearance: White to off-white
MDL No.: MFCD00150104
Excellent Colourimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result
Formel:
C₁₆H₂₀N₂·2HCl·xH₂O Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD00150104 CAS-Nummer: 207738-08-7 EINECS: 264-769-6 |