Dextransulfat Natriumsalz, M.W. ~ 40,000
Lieferant: Thermo Fisher Scientific
Synonyme:
Dextran sulphate sodium from Dextran
J63606.14
J63606.22
J63606.PG
J63606.14EA
83.9
EUR
J63606.14
J63606.22
J63606.PG
Dextransulfat Natriumsalz, M.W. ~ 40,000
Dextran sulphate sodium salt
CAS No.: 9011-18-1
Physical Form: Powder
Appearance: Off-white to pale yellow
Odour: Phenol-like
Merck Reference: 14,2951
MDL No.: MFCD00081551
Formel:
(C₆H₇Na₃O₁₄S₃)n Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00081551 CAS-Nummer: 9011-18-1 Merck Index: 14,2951 |