Erythrosin B, Pulver, in Spiritus löslich

Lieferant: Thermo Fisher Scientific

Synonyme: Tetraiodofluorescein, Solvent Red 140, C.I. 45430:2

J62619.06 J62619.14 J62619.22
J62619.06EA 34.5 EUR
J62619.06 J62619.14 J62619.22
Erythrosin B, Pulver, in Spiritus löslich
Erythrosin B
Formel: C₂₀H₈I₄O₅
Molekulargewicht: 835.89 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00005044
CAS-Nummer: 15905-32-5
EINECS: 240-046-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR