Polymyxin B-Sulfat, Zellkultur-Qualität Cell Culture Reagent

Lieferant: Thermo Fisher Scientific

J61763.03 J61763.06
J61763.03EA 115 EUR
J61763.03 J61763.06
Polymyxin B-Sulfat, Zellkultur-Qualität Cell Culture Reagent
Polymyxin B sulphate

Antibiotic for Gram-negative bacteria


  • White to pale yellow powder


Soluble in water

Formel: C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00131991
CAS-Nummer: 1405-20-5
Merck Index: 14,7573

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR