L(+)-Lysin Monohydrochlorid Reagenz für die Zellkultur
Lieferant: Thermo Fisher Scientific
Synonyme:
Lysine Hydrochloride, (S)-(+)-2,6-Diaminohexanoic acid monohydrochloride, L-Lysine monohydrochloride, H-Lys-OH·HCl, L-Lysine monohydrocholoride, L-Lysine Hydrochloride, (S)-(+)-Lysine monohydrochloride, (S)-(+)-2,6-Diaminocaproic acid monohydrochloride, H-Lys-OH.HCl
CAS No.: 657-27-2
Molecular Formula: C6H14N2O2.HCl
Formula Weight: 182.65
Physical Form: Powder
Appearance: White to off-white
Beilstein Reference: 3563889
Merck Reference: 14,5636
MDL No.: MFCD00064564
Formel:
H₂N(CH₂)₄CH(NH₂)CO₂H·HCl Schmelzpunkt: 263…264 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00064564 CAS-Nummer: 657-27-2 EINECS: 211-519-9 Merck Index: 14,5636 |