Pyronin G

Lieferant: Thermo Fisher Scientific

Synonyme: Pyronin Y, Pyronine Y, 3,6-Bis(dimethylamino)xanthylium chloride

J61068.03 J61068.06 J61068.09
J61068.03EA 38.5 EUR
J61068.03 J61068.06 J61068.09
Pyronin G
Pyronin G

CAS No.: 92-32-0 Molecular Formula: C17H19ClN2O Formula Weight: 302.80 Physical Form: Crystalline powder Appearance: Green Melting Point: 250-260° Beilstein Reference: 3795789 Merck Reference: 14,8007 MDL No.: MFCD00011725


Marker dye for RNA useful in acid buffer systems

Formel: C₁₇H₁₉ClN₂O
Molekulargewicht: 302.8 g/mol
Schmelzpunkt: 250…260 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00011725
CAS-Nummer: 92-32-0
EINECS: 202-147-8
Merck Index: 12,08190

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR