myo-Inosit ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme:
i-Inositol, D-Myo-Inositol, rel-(1R,2r,3S,4R,5s,6S)-Cyclohexane-1,2,3,4,5,6-hexaol, Inositol
Purity: 99%
CAS No.: 87-89-8
Molecular Formula: C6H12O6
Formula Weight: 180.16
Physical Form: Crystalline powder
Appearance: White
Odour: Odourless
Melting Point: 222-227°
Beilstein Reference: 1907329
Merck Reference: 14,4978
MDL No.: MFCD00077932
A lipotropic agent
Formel:
C₆H₁₂O₆ Molekulargewicht: 180.16 g/mol Siedepunkt: 291 Schmelzpunkt: 222…227 °C Dichte: 1.752 g/cm³ (20 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00077932 CAS-Nummer: 87-89-8 EINECS: 201-781-2 Merck Index: 13,05001 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...