Ammoniumsulfat 99+%, ultrarein
Lieferant: Thermo Fisher Scientific
Achtung
|
Synonyme:
Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite
J64419.22
J64419.36
J64419.A3
J64419.22EA
27.9
EUR
J64419.22
J64419.36
J64419.A3
Ammoniumsulfat 99+%, ultrarein
Ammoniumsulfat
Purity: 99+%
CAS No.: 7783-20-2
Molecular Formula: H8N2O4S
Formula Weight: 132.14
Physical Form: Crystalline
Density: 1.769
Appearance: White
Melting Point: ca 280° dec.
Merck Reference: 14,555
MDL No.: MFCD00003391
Formel:
(NH₄)₂SO₄ Schmelzpunkt: ∼ 280 °C (dec.) Dichte: 1.769 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00003391 CAS-Nummer: 7783-20-2 EINECS: 231-984-1 REACH-Nummer: 01-2119455044-46 Merck Index: 14,555 |