Lanthan(III)carbonat Hydrat ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Lieferant: Thermo Fisher Scientific
|
Achtung
|
13197.18
13197.30
13197.18EA
114
EUR
13197.18
13197.30
Lanthan(III)carbonat Hydrat ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Lanthan(III)carbonat Hydrat
Purity: 99.99%
CAS No.: 54451-24-0
Molecular Formula: La2(CO3)3.xH2O
Formula Weight: 457.85(anhy)
Physical Form: Powder/Lump
Density: 2.6-2.7
Merck Reference: 14,5364
MDL No.: MFCD00011069
Formel:
C₃La₂O₉·H₂O Molekulargewicht: 457.85 g/mol (anhy) Dichte: 2.6…2.7 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011069 CAS-Nummer: 54451-24-0 EINECS: 209-599-5 Merck Index: 14,5364 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...