Cer(III)carbonat Hydrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

325503-250G 325503-50G
SIAL325503-250GEA 233 EUR
SIAL325503-250G SIAL325503-50G
Cer(III)carbonat Hydrat, Sigma-Aldrich®
Cer(III)carbonat Hydrat
Formel: C₃Ce₂O₉·H₂O
MDL Number: MFCD00149635
CAS-Nummer: 54451-25-1
EINECS: 208-655-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR