Nickel(II)sulfamat Hydrat
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
89020.30
89020.18
89020.A1
89020.30EA
224
EUR
89020.30
89020.18
89020.A1
Nickel(II)sulfamat Hydrat
Nickel(II)sulfamat Hydrat
Nickel(II) sulfamate is used as an electro plating and surface treating agent. It finds application in metal coloring and casting.
CAS No.: 13770-89-3
Molecular Formula: Ni(SO3NH2)2.xH2O
Formula Weight: 250.86(anhy)
Physical Form: Crystalline
Appearance: Blue
Odour: Odourless
UN No.: 3077
MDL No.: MFCD00137261
Formel:
H₄N₂NiO₆S₂·H₂O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00137261 CAS-Nummer: 13770-89-3 EINECS: 237-396-1 UN-Nummer: 3077 ADR: 9,III |
Spezifikation Testergebnisse
Assay (unspecified) | 15.1%-19.2% Ni |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...