6-Methylchromon ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
6-Methyl-4H-chromen-4-one
H27002.06
H27002.03
H27002.06EA
147
EUR
H27002.06
H27002.03
6-Methylchromon ≥98%
6-Methylchromon
Purity: 98%
CAS No.: 38445-23-7
Molecular Formula: C10H8O2
Formula Weight: 160.17
Melting Point: 88-90°
MDL No.: MFCD00218598
Formel:
C₁₀H₈O₂ Schmelzpunkt: 88…90 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00218598 CAS-Nummer: 38445-23-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...