4'-(4-Bromphenyl)acetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Acetyl-4'-bromobiphenyl, 1-(4'-Bromo-[1,1'-biphenyl]-4-yl)ethanone

640468-100G 640468-25G
SIAL640468-100GEA 164 EUR
SIAL640468-100G SIAL640468-25G
4'-(4-Bromphenyl)acetophenon, Sigma-Aldrich®
4'-(4-Bromphenyl)acetophenon
Formel: C₁₄H₁₁BrO
MDL Number: MFCD00143242
CAS-Nummer: 5731-01-1
EINECS: 227-236-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR