4-(2-Carboxyethyl)phenylboronsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-(2-Carboxyethyl)benzeneboronic acid

L17485.03 L17485.06
L17485.03EA 106 EUR
L17485.03 L17485.06
4-(2-Carboxyethyl)phenylboronsäure ≥97%
4-(2-Carboxyethyl)phenylboronsäure

Purity: 97% CAS No.: 166316-48-9 Molecular Formula: C9H11BO4 Formula Weight: 193.99 Melting Point: 146-148° Beilstein Reference: 9198524

Formel: C₉H₁₁BO₄
Schmelzpunkt: 146…148 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01318119
CAS-Nummer: 166316-48-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR